Gene Transcripts
- Gene Symbol
- SLC38A9
- Entrez Gene ID
- 153129
- Gene Name
- solute carrier family 38 member 9
- Gene Aliases
- URLC11
- Chromosome Location
- Chr. 5: 55625845 - 55712344 on Build GRCh38
- UniGene ID
- Hs.742101
- Species
- Homo sapiens
View Full Product Details