Gene Transcripts

Gene Symbol
RUSC1-AS1
Entrez Gene ID
284618
Gene Name
RUSC1 antisense RNA 1
Gene Aliases
C1orf104
Chromosome Location
Chr. 1: 155320460 - 155324147 on Build GRCh38
UniGene ID
Hs.650801
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK093295.1 - 4 795
AK125510.1 - 1 2426
BC131614.1 - 2 884 40115930

View Full Product Details