Gene Transcripts

Gene Symbol
EMC10
Entrez Gene ID
284361
Gene Name
ER membrane protein complex subunit 10
Gene Aliases
C19orf63, HSM1, HSS1
Chromosome Location
Chr. 19: 50476477 - 50505905 on Build GRCh38
UniGene ID
Hs.448941
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175063.5 NP_778233.4 6 727
NM_206538.3 NP_996261.1 6 727
XM_006723163.3 XP_006723226.1 6 721
XM_011526818.2 XP_011525120.1 6 721
GenBank mRNA AL512761.1 - 2 243
AY194293.2 - 10 1685
AY358710.1 - 6 687
AY761095.1 - 6 705
AY761097.1 - 6 705
BC032948.2 - 6 668 4794992
BC035001.2 - 6 666 4796476
BC062607.1 - 6 869
CN423491.1 - 3 352
HY149334.1 - 3 361

View Full Product Details