Gene Transcripts
- Gene Symbol
- DGCR6
- Entrez Gene ID
- 8214
- Gene Name
- DiGeorge syndrome critical region gene 6
- Gene Aliases
- Chromosome Location
- Chr. 22: 18906223 - 18912088 on Build GRCh38
- UniGene ID
- Hs.474185
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
X96484.1 |
- |
|
1064 |
|
View Full Product Details