Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016417.2 | NP_057501.2 | 2 | 632 | |
GenBank mRNA | AB223038.1 | - | 2 | 571 | |
AF113691.1 | - | 202 | |||
BC023528.2 | - | 2 | 582 | 3613760 | |
BC047680.1 | - | 2 | 578 | 6066312 | |
BC105275.1 | - | 1 | 109 | ||
BI834047.1 | - | 2 | 380 | ||
BX248075.1 | - | 2 | 585 |