Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278503.1 | NP_001265432.1 | 4 | 383 | |
NM_001278504.1 | NP_001265433.1 | 2 | 172 | ||
NM_152713.4 | NP_689926.1 | 3 | 295 | ||
XM_011542807.2 | XP_011541109.1 | 3 | 322 | ||
GenBank mRNA | AK290040.1 | - | 3 | 245 | |
AK290657.1 | - | 3 | 243 | ||
AK295892.1 | - | 2 | 100 | ||
AK308902.1 | - | 3 | 238 | ||
BC020965.2 | - | 3 | 228 | 3891543 | |
BC048348.1 | - | 2 | 181 | 5561615 | |
BT007100.1 | - | 2 | 136 | ||
BX649102.1 | - | 4 | 321 | ||
DA577274.1 | - | 3 | 295 | ||
L38961.1 | - | 3 | 242 | ||
L47337.1 | - | 3 | 252 |