Gene Transcripts

Gene Symbol
FAAP100
Entrez Gene ID
80233
Gene Name
Fanconi anemia core complex associated protein 100
Gene Aliases
C17orf70
Chromosome Location
Chr. 17: 81539885 - 81552403 on Build GRCh38
UniGene ID
Hs.313905
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025161.5 NP_079437.5 6 2329
NR_033338.1 6 2525
XM_006722111.2 XP_006722174.1 6 1910
XM_006722112.3 XP_006722175.1 6 2475
XM_017025162.1 XP_016880651.1 6 2350
XM_017025163.1 XP_016880652.1 6 2246
GenBank mRNA AK024644.1 - 5 756
AK025828.1 - 2 590
AK054713.1 - 2 1673
AK225370.1 - 2 590
AL834374.1 - 2 814
BC001724.1 - 1 88
BC008883.2 - 6 2498
BC021968.2 - 2 401
BC117139.1 - 4 1849 40125690
BC117141.1 - 4 1849 40125692
BC143861.1 - 4 1849
DQ989324.1 - 6 2311

View Full Product Details