Gene Transcripts
- Gene Symbol
- WDR83OS
- Entrez Gene ID
- 51398
- Gene Name
- WD repeat domain 83 opposite strand
- Gene Aliases
- C19orf56, PTD008
- Chromosome Location
- Chr. 19: 12668067 - 12671023 on Build GRCh38
- UniGene ID
- Hs.108969
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016145.3 |
NP_057229.1
|
3, 4 |
501 |
|
XM_017026864.1 |
XP_016882353.1
|
4, 5 |
1495 |
|
GenBank mRNA |
AF059620.1 |
- |
3, 4 |
269 |
|
AF078861.1 |
- |
3, 4 |
486 |
|
AF151898.1 |
- |
3, 4 |
485 |
|
AK091030.1 |
- |
3, 4 |
2206 |
|
AK311944.1 |
- |
3, 4 |
265 |
|
BC001192.1 |
- |
3, 4 |
249 |
|
BC042919.1 |
- |
3, 4 |
270 |
5932109 |
BC061912.1 |
- |
3, 4 |
270 |
4444565 |
View Full Product Details