Gene Transcripts

Gene Symbol
Mpst
Entrez Gene ID
192172
Gene Name
mercaptopyruvate sulfurtransferase
Gene Aliases
Mst
Chromosome Location
Chr. 7: 119626636 - 119631317 on Build Rnor_6.0
UniGene ID
Rn.32263
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138843.1 NP_620198.1 2 842
GenBank mRNA BC086575.1 - 2 657
D50564.1 - 2 842

View Full Product Details