Gene Transcripts

Gene Symbol
Serpinf1
Entrez Gene ID
287526
Gene Name
serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 1
Gene Aliases
Dmrs91, Pedf
Chromosome Location
Chr. 10: 62241750 - 62254145 on Build Rnor_6.0
UniGene ID
Rn.16993
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177927.2 NP_808788.1 3 218
GenBank mRNA AY216659.1 - 3 218
BC078686.1 - 3 206 7110230
FQ218413.1 - 3 218
FQ219756.1 - 3 217

View Full Product Details