Gene Transcripts

Gene Symbol
Maob
Entrez Gene ID
25750
Gene Name
monoamine oxidase B
Gene Aliases
Chromosome Location
Chr. X: 6430694 - 6533520 on Build Rnor_6.0
UniGene ID
Rn.6656
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013198.1 NP_037330.1 2 192
GenBank mRNA BC078886.1 - 2 199
BC089814.1 - 2 157 7323595
FQ210326.1 - 2 253
M23601.1 - 2 192

View Full Product Details