Gene Transcripts

Gene Symbol
Nps
Entrez Gene ID
100043254
Gene Name
neuropeptide S
Gene Aliases
ENSMUSG00000073804
Chromosome Location
Chr. 7: 135268619 - 135272942 on Build GRCm38
UniGene ID
Mm.192092
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163611.1 NP_001157083.1 3 839

View Full Product Details