Gene Transcripts

Gene Symbol
Zfc3h1
Entrez Gene ID
216345
Gene Name
zinc finger, C3H1-type containing
Gene Aliases
BC033596, Ccdc131, Psrc2
Chromosome Location
Chr. 10: 115384920 - 115432772 on Build GRCm38
UniGene ID
Mm.207621
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033261.2 NP_001028433.2 12 2709
GenBank mRNA AK145652.1 - 12 2711
AK172969.1 - 10 1409
BC139124.1 - 12 2566 8862146

View Full Product Details