Gene Transcripts

Gene Symbol
Aldh1a7
Entrez Gene ID
29651
Gene Name
aldehyde dehydrogenase family 1, subfamily A7
Gene Aliases
Aldh1a4, Aldhpb
Chromosome Location
Chr. 1: 240561951 - 240601828 on Build Rnor_6.0
UniGene ID
Rn.74044
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017272.15 NP_058968.14 7 776
XM_008760315.1 XP_008758537.1 7 800
GenBank mRNA M23995.1 - 7 776

View Full Product Details