Gene Transcripts

Gene Symbol
Magea10
Entrez Gene ID
236852
Gene Name
melanoma antigen family A, 10
Gene Aliases
Chromosome Location
Chr. X: 72381870 - 72386858 on Build GRCm38
UniGene ID
Mm.115316
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085506.1 NP_001078975.1 4 1375
GenBank mRNA AK136064.1 - 4 1376

View Full Product Details