Gene Transcripts

Gene Symbol
Skint5
Entrez Gene ID
242627
Gene Name
selection and upkeep of intraepithelial T cells 5
Gene Aliases
OTTMUSG00000008560, Skint-5
Chromosome Location
Chr. 4: 113477860 - 113999506 on Build GRCm38
UniGene ID
Mm.484115
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167876.1 NP_001161348.1 55 3997
NM_001167878.1 NP_001161350.1 54 3934
GenBank mRNA EU099301.1 - 55 3997
EU099302.1 - 54 3934

View Full Product Details