Gene Transcripts

Gene Symbol
Layn
Entrez Gene ID
244864
Gene Name
layilin
Gene Aliases
E030012M19Rik, Gm511
Chromosome Location
Chr. 9: 51056780 - 51077209 on Build GRCm38
UniGene ID
Mm.335152
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033534.1 NP_001028706.1 3 498
GenBank mRNA AK086930.1 - 3 459
AK165695.1 - 3 452
BC139816.1 - 470 40140447

View Full Product Details