Gene Transcripts

Gene Symbol
Lmod3
Entrez Gene ID
320502
Gene Name
leiomodin 3 (fetal)
Gene Aliases
5430424A14Rik
Chromosome Location
Chr. 6: 97238528 - 97253097 on Build GRCm38
UniGene ID
Mm.68163
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081157.1 NP_001074626.1 2 1410
GenBank mRNA AK030676.1 - 2 1392
BC127157.1 - 2 1338 40091077

View Full Product Details