Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001033409.3 | NP_001028581.1 | 18 | 2876 | |
GenBank mRNA | AK040883.1 | - | 1 | 478 | |
AK052873.1 | - | 13 | 2077 | ||
AK085901.1 | - | 18 | 2838 | ||
AK137380.1 | - | 18 | 2877 | ||
BC026896.1 | - | 3 | 1218 | ||
BC141210.1 | - | 18 | 2828 | 9055822 |