Gene Transcripts

Gene Symbol
Tmcc1
Entrez Gene ID
330401
Gene Name
transmembrane and coiled coil domains 1
Gene Aliases
3632431M01Rik, mKIAA0779
Chromosome Location
Chr. 6: 116018611 - 116193379 on Build GRCm38
UniGene ID
Mm.425352
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177412.1 NP_803131.1 6 3649
GenBank mRNA AK036701.1 - 1 1724
AK173022.1 - 6 3638
BC049661.1 - 1 92

View Full Product Details