Gene Transcripts

Gene Symbol
Siah3
Entrez Gene ID
380918
Gene Name
seven in absentia homolog 3 (Drosophila)
Gene Aliases
Gm912
Chromosome Location
Chr. 14: 75455982 - 75526141 on Build GRCm38
UniGene ID
Mm.484360
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128093.1 NP_001121565.1 2 829
GenBank mRNA BC147762.1 - 2 829
BC147764.1 - 2 829 9087489

View Full Product Details