Gene Transcripts

Gene Symbol
Vmn1r39
Entrez Gene ID
546903
Gene Name
vomeronasal 1 receptor 39
Gene Aliases
EG546903, Gm5993
Chromosome Location
Chr. 6: 66804415 - 66805332 on Build GRCm38
UniGene ID
Mm.484975
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166720.1 NP_001160192.1 1 864

View Full Product Details