Gene Transcripts

Gene Symbol
Vmn1r61
Entrez Gene ID
636731
Gene Name
vomeronasal 1 receptor 61
Gene Aliases
Gm7186
Chromosome Location
Chr. 7: 5610411 - 5611313 on Build GRCm38
UniGene ID
Mm.479999
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166733.1 NP_001160205.1 1 829

View Full Product Details