Gene Transcripts

Gene Symbol
Pcp4l1
Entrez Gene ID
66425
Gene Name
Purkinje cell protein 4-like 1
Gene Aliases
1500036F01Rik
Chromosome Location
Chr. 1: 171173264 - 171196268 on Build GRCm38
UniGene ID
Mm.482117
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025557.1 NP_079833.1 3 1521
GenBank mRNA AK002772.1 - 3 1492
AK075738.1 - 3 1491
AY304481.1 - 3 1492

View Full Product Details