Gene Transcripts

Gene Symbol
Krt28
Entrez Gene ID
70843
Gene Name
keratin 28
Gene Aliases
4733401L19Rik, Krt25d
Chromosome Location
Chr. 11: 99364861 - 99379930 on Build GRCm38
UniGene ID
Mm.180256
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027574.1 NP_081850.1 1 277
GenBank mRNA AB288231.1 - 1 277
AK014642.1 - 1 278

View Full Product Details