Gene Transcripts

Gene Symbol
Spef1
Entrez Gene ID
70997
Gene Name
sperm flagellar 1
Gene Aliases
4931426K16Rik, Clamp
Chromosome Location
Chr. 2: 131170261 - 131174810 on Build GRCm38
UniGene ID
Mm.252722
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027641.2 NP_081917.1 7 2242
GenBank mRNA AK046964.1 - 2308

View Full Product Details