Gene Transcripts

Gene Symbol
Glt6d1
Entrez Gene ID
71103
Gene Name
glycosyltransferase 6 domain containing 1
Gene Aliases
4933411C14Rik, GT6m7
Chromosome Location
Chr. 2: 25771820 - 25815940 on Build GRCm38
UniGene ID
Mm.236431
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039095.1 NP_001034184.1 5 717
GenBank mRNA AK016765.1 - 5 724
BC111823.1 - 5 717 40052059

View Full Product Details