Gene Transcripts

Gene Symbol
Enkur
Entrez Gene ID
71233
Gene Name
enkurin, TRPC channel interacting protein
Gene Aliases
4933434I06Rik, AU016229
Chromosome Location
Chr. 2: 21180731 - 21205365 on Build GRCm38
UniGene ID
Mm.27658
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027728.2 NP_082004.1 1 117
GenBank mRNA AK017056.1 - 1 118
AY454124.1 - 1 115
BC099519.1 - 1 16 30848009

View Full Product Details