Gene Transcripts

Gene Symbol
Enkur
Entrez Gene ID
71233
Gene Name
enkurin, TRPC channel interacting protein
Gene Aliases
4933434I06Rik, AU016229
Chromosome Location
Chr. 2: 21180731 - 21205365 on Build GRCm38
UniGene ID
Mm.27658
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027728.2 NP_082004.1 1, 2 294
GenBank mRNA AK017056.1 - 1, 2 295
AY454124.1 - 1, 2 292
BC099519.1 - 1, 2 193 30848009
BC132425.1 - 1, 2 155 40130702
BC132451.1 - 1, 2 155 40130728

View Full Product Details