Gene Transcripts

Gene Symbol
Ttc25
Entrez Gene ID
74407
Gene Name
tetratricopeptide repeat domain 25
Gene Aliases
4933404O19Rik
Chromosome Location
Chr. 11: 100545613 - 100572566 on Build GRCm38
UniGene ID
Mm.31590
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028918.2 NP_083194.2 3 484
GenBank mRNA AK016657.1 - 3 415
AK161541.1 - 3 423
AK162855.1 - 3 424
BC050805.1 - 3 508 6771545
CA463990.1 - 3 511

View Full Product Details