Gene Transcripts

Gene Symbol
Fam46c
Entrez Gene ID
74645
Gene Name
family with sequence similarity 46, member C
Gene Aliases
4930431B09Rik, AI449797
Chromosome Location
Chr. 3: 100468057 - 100489192 on Build GRCm38
UniGene ID
Mm.226900
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142952.1 NP_001136424.1 2 1543
GenBank mRNA AK015259.1 - 1 1347
BC037072.1 - 2 1533
BC046309.1 - 2 1537

View Full Product Details