Gene Transcripts

Gene Symbol
Ephx2
Entrez Gene ID
65030
Gene Name
epoxide hydrolase 2, cytoplasmic
Gene Aliases
Chromosome Location
Chr. 15: 42757241 - 42794263 on Build Rnor_6.0
UniGene ID
Rn.54495
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022936.1 NP_075225.1 2 215
GenBank mRNA BC085732.1 - 2 220
X65083.1 - 2 215

View Full Product Details