Gene Transcripts

Gene Symbol
Prx
Entrez Gene ID
78960
Gene Name
periaxin
Gene Aliases
Chromosome Location
Chr. 1: 84302912 - 84324560 on Build Rnor_6.0
UniGene ID
Rn.10395
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023976.2 NP_076466.2 6 553
XM_006228586.2 XP_006228648.1 6 646
XM_008759196.1 XP_008757418.1 5 747
GenBank mRNA Z29649.2 - 6 553

View Full Product Details