Gene Transcripts

Gene Symbol
Gabbr2
Entrez Gene ID
83633
Gene Name
gamma-aminobutyric acid (GABA) B receptor 2
Gene Aliases
Gpr51
Chromosome Location
Chr. 5: 62276100 - 62621737 on Build Rnor_6.0
UniGene ID
Rn.162814
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031802.1 NP_113990.1 3, 4 785
GenBank mRNA AF058795.1 - 3, 4 785
AF074482.1 - 3, 4 635
AF109405.1 - 3, 4 752

View Full Product Details