Gene Transcripts
- Gene Symbol
- Zp1
- Entrez Gene ID
- 85271
- Gene Name
- zona pellucida glycoprotein 1 (sperm receptor)
- Gene Aliases
- Chromosome Location
- Chr. 1: 226966366 - 226972746 on Build Rnor_6.0
- UniGene ID
- Rn.10890
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053509.1 |
NP_445961.1
|
9, 10 |
1588 |
|
GenBank mRNA |
AB000928.1 |
- |
9, 10 |
1588 |
|
View Full Product Details