Gene Transcripts

Gene Symbol
Dll3
Entrez Gene ID
114125
Gene Name
delta-like 3 (Drosophila)
Gene Aliases
Chromosome Location
Chr. 1: 85485866 - 85493683 on Build Rnor_6.0
UniGene ID
Rn.23105
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053666.1 NP_446118.1 6 1071
XM_006228611.2 XP_006228673.1 6 1130
GenBank mRNA AF084576.1 - 6 1071

View Full Product Details