Gene Transcripts

Gene Symbol
Arl5a
Entrez Gene ID
117050
Gene Name
ADP-ribosylation factor-like 5A
Gene Aliases
Arl5
Chromosome Location
Chr. 3: 37922544 - 37947434 on Build Rnor_6.0
UniGene ID
Rn.2803
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053979.2 NP_446431.1 6 701
XM_008761696.1 XP_008759918.1 6 773
GenBank mRNA BC097294.1 - 6 701
X78604.1 - 6 550

View Full Product Details