Gene Transcripts

Gene Symbol
Ugt1a7c
Entrez Gene ID
154516
Gene Name
UDP glucuronosyltransferase 1 family, polypeptide A7C
Gene Aliases
UDPGT 1-7, Ugt1, Ugt1a7, Ugt1a8
Chromosome Location
Chr. 9: 95233957 - 95302822 on Build Rnor_6.0
UniGene ID
Rn.26489
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130407.2 NP_569091.2 293
GenBank mRNA AF461738.1 - 301
AY435134.1 - 260
BC078732.1 - 296
U75903.1 - 302

View Full Product Details