Gene Transcripts
- Gene Symbol
- Sep15
- Entrez Gene ID
- 113922
- Gene Name
- selenoprotein 15
- Gene Aliases
- Chromosome Location
- Chr. 2: 250600823 - 250633903 on Build Rnor_6.0
- UniGene ID
- Rn.1124
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_133297.2 |
NP_579831.2
|
4, 5 |
627 |
|
GenBank mRNA |
AF390544.1 |
- |
4, 5 |
351 |
|
BC060547.1 |
- |
4, 5 |
376 |
6921948 |
CK596377.1 |
- |
4, 5 |
642 |
|
DY561321.1 |
- |
4, 5 |
365 |
|
FQ209574.1 |
- |
4, 5 |
362 |
|
FQ212727.1 |
- |
4, 5 |
395 |
|
FQ213177.1 |
- |
4, 5 |
376 |
|
FQ213838.1 |
- |
4, 5 |
362 |
|
FQ214634.1 |
- |
4, 5 |
372 |
|
FQ215362.1 |
- |
4, 5 |
362 |
|
FQ215992.1 |
- |
4, 5 |
385 |
|
FQ216831.1 |
- |
4, 5 |
362 |
|
FQ219237.1 |
- |
4, 5 |
383 |
|
FQ219570.1 |
- |
4, 5 |
362 |
|
FQ219725.1 |
- |
4, 5 |
355 |
|
FQ220593.1 |
- |
4, 5 |
367 |
|
FQ220877.1 |
- |
4, 5 |
375 |
|
FQ229133.1 |
- |
4, 5 |
368 |
|
FQ230131.1 |
- |
4, 5 |
362 |
|
View Full Product Details