Gene Transcripts

Gene Symbol
Pacs1
Entrez Gene ID
171444
Gene Name
phosphofurin acidic cluster sorting protein 1
Gene Aliases
Pacs-1
Chromosome Location
Chr. 1: 220515117 - 220644825 on Build Rnor_6.0
UniGene ID
Rn.154425
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134406.1 NP_599233.1 3 557
GenBank mRNA AF076183.1 - 3 557
AF076184.1 - 3 557
HM537135.1 - 3 352

View Full Product Details