Gene Transcripts

Gene Symbol
Myo16
Entrez Gene ID
192253
Gene Name
myosin XVI
Gene Aliases
Myr8
Chromosome Location
Chr. 16: 84575149 - 84963589 on Build Rnor_6.0
UniGene ID
Rn.207204
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138893.1 NP_620248.1 1 242
XM_008771384.1 XP_008769606.1 2 1216
GenBank mRNA AF209114.1 - 1 242
AY004215.1 - 1 242

View Full Product Details