Gene Transcripts

Gene Symbol
Gja4
Entrez Gene ID
25655
Gene Name
gap junction protein, alpha 4
Gene Aliases
CXN37
Chromosome Location
Chr. 5: 145374688 - 145377221 on Build Rnor_6.0
UniGene ID
Rn.23406
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021654.2 NP_067686.1 2 328
GenBank mRNA BC078837.1 - 2 328
BC086576.1 - 1 259 7306623

View Full Product Details