Gene Transcripts
- Gene Symbol
- Htr1b
- Entrez Gene ID
- 25075
- Gene Name
- 5-hydroxytryptamine (serotonin) receptor 1B, G protein-coupled
- Gene Aliases
- Chromosome Location
- Chr. 8: 89113997 - 89130829 on Build Rnor_6.0
- UniGene ID
- Rn.138109
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022225.1 |
NP_071561.1
|
1 |
144 |
|
GenBank mRNA |
X62944.1 |
- |
1 |
180 |
|
View Full Product Details