Gene Transcripts

Gene Symbol
Ceacam10
Entrez Gene ID
26366
Gene Name
carcinoembryonic antigen-related cell adhesion molecule 10
Gene Aliases
Bgp3, Cea10
Chromosome Location
Chr. 7: 24777156 - 24784656 on Build GRCm38
UniGene ID
Mm.30300
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007675.4 NP_031701.3 2 431
GenBank mRNA AK018529.1 - 2 411
BC003346.1 - 2 452 3481822
L38422.1 - 2 395

View Full Product Details