Gene Transcripts

Gene Symbol
Pla2g2c
Entrez Gene ID
18781
Gene Name
phospholipase A2, group IIC
Gene Aliases
Chromosome Location
Chr. 4: 138725325 - 138744575 on Build GRCm38
UniGene ID
Mm.5189
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008868.3 NP_032894.2 4 440
GenBank mRNA BC029347.1 - 3 285
CA464794.1 - 4 443
U18119.1 - 3 266

View Full Product Details