Gene Transcripts

Gene Symbol
Pgr
Entrez Gene ID
18667
Gene Name
progesterone receptor
Gene Aliases
9930019P03Rik, BB114106, ENSMUSG00000074510, NR3C3, PR, PR-A, PR-B
Chromosome Location
Chr. 9: 8894736 - 8968611 on Build GRCm38
UniGene ID
Mm.12798
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008829.2 NP_032855.2 5 2919
GenBank mRNA AK054117.1 - 5 1600
BC145807.1 - 5 2324 40131110
M68915.1 - 5 2282

View Full Product Details