Gene Transcripts

Gene Symbol
Slc50a1
Entrez Gene ID
19729
Gene Name
solute carrier family 50 (sugar transporter), member 1
Gene Aliases
MmSWEET1, Rag1ap1, Rga
Chromosome Location
Chr. 3: 89268246 - 89270570 on Build GRCm38
UniGene ID
Mm.17958
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009057.3 NP_033083.2 6 955
GenBank mRNA AK014305.1 - 6 908
AK045924.1 - 4 1268
AK079716.1 - 5 1020
AK158862.1 - 5 980
AK170162.1 - 6 906
AK209820.1 - 1 202
BC014292.1 - 6 939 4190124
X96618.1 - 6 946

View Full Product Details