Gene Transcripts

Gene Symbol
Usf1
Entrez Gene ID
22278
Gene Name
upstream transcription factor 1
Gene Aliases
bHLHb11
Chromosome Location
Chr. 1: 171411304 - 171418759 on Build GRCm38
UniGene ID
Mm.8
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK142437.1 - 4, 5 381
BC049784.1 - 4, 5 324 6506829
CJ050979.1 - 4, 5 460

View Full Product Details