Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001277268.1 | NP_001264197.1 | 2 | 1040 | |
NM_010203.5 | NP_034333.1 | 3 | 1144 | ||
GenBank mRNA | AK028694.1 | - | 3 | 1001 | |
AK028894.1 | - | 3 | 1146 | ||
BC071227.1 | - | 3 | 1117 | 30443152 |