Gene Transcripts

Gene Symbol
Gzme
Entrez Gene ID
14942
Gene Name
granzyme E
Gene Aliases
CCP3, Ctla-6, Ctla6, MCSP-2
Chromosome Location
Chr. 14: 56117619 - 56120625 on Build GRCm38
UniGene ID
Mm.14424
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010373.3 NP_034503.2 2, 3 277
GenBank mRNA BG146331.1 - 2, 3 317
J03256.1 - 2, 3 189
M36901.1 - 2, 3 169
X14093.1 - 2, 3 169

View Full Product Details